Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl CoA synthetase Antibody (OTI3H4), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272039H
Description
Acetyl CoA synthetase Monoclonal specifically detects Acetyl CoA synthetase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Acetyl CoA synthetase | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ACSS2 | |
Full length recombinant protein of human Acetyl CoA synthetase (NP_061147) produced in HEK293T cells. | |
0.1 mL | |
55902.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI3H4 | |
Western Blot, Immunohistochemistry | |
ACAS2, AceCS, acetate thiokinase, acetate-CoA ligase, Acetate--CoA ligase, Acetyl-CoA synthetase, acetyl-Coenzyme A synthetase 2 (ADP forming), acetyl-coenzyme A synthetase, cytoplasmic, ACSA, ACSACECS, Acyl-activating enzyme, acyl-CoA synthetase short-chain family member 2EC 6.2.1.1, cytoplasmic acetyl-coenzyme A synthetase, dJ1161H23.1, DKFZp762G026, EC 6.2.1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction