Learn More
Acetyl Cortactin (Lys309) Rabbit anti-Monkey, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Acetyl Cortactin (Lys309) |
| Applications | Immunofluorescence, Western Blot |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | CTTN,EMS1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 14 amino acids surrounding acetylated lysine309 of human Src substrate cortactin. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.