Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acetyl-HIST1H2BB (Lys5) Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144284
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144284 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144284 Supplier Invitrogen™ Supplier No. PA5144284
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acetyl-HIST1H2BB (Lys5) Polyclonal Antibody for ICC/IF, ELISA, ChIP

Core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones thereby play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA accessibility is regulated via a complex set of post-translational modifications of histones, also called histone code, and nucleosome remodeling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl-HIST1H2BB (Lys5)
Applications ChIP Assay, ELISA, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene H2BC3
Gene Accession No. P33778
Gene Alias H2B histone family, member F; H2B.1; H2B/f; h2B-143; H2B-clustered histone 3; H2BFF; HIST1H2BB; histone 1, H2bb; histone cluster 1, H2bb; histone H2B type 1-B; Histone H2B.1; histone H2B.f
Gene Symbols H2BC3, HIST1H2BB
Host Species Rabbit
Immunogen Peptide sequence around site of Acetyl-Lys (5) derived from Human Histone H2B type 1-B.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3018
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.