Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H2A.Zac pan-acetyl (K4,K7,K11) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540095
Description
H2A.Zac pan-acetyl (K4,K7,K11) Polyclonal Antibody for ICC/IF, ChIP, ChIP-seq, ELISA
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.
Specifications
H2A.Zac pan-acetyl (K4,K7,K11) | |
Polyclonal | |
Unconjugated | |
H2AZ1 | |
11ac; Acetyl-Histone H2A.Z K4 K7 K11; Acetyl-Histone H2A.Z Lys4 Lys7 Lys11; H2A histone family member Z; H2A histone family, member Z; H2A.z; H2A.Z-1; H2A.ZK4,7,11ac; H2A.ZK5; H2A.ZK5ac; H2A.ZLys4,7,11ac; H2A/z; H2AFZ; H2AFZac; H2AZ; H2AZ histone; H2AZK11ac; H2AZK4,7,11ac; H2AZK7ac; H2AZLys4,7,11ac; histone H2A.Z | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3015, 94239 | |
-20°C or -80°C if preferred | |
Liquid |
ChIP Assay, ChIP sequencing (ChIP-seq), ELISA, Immunocytochemistry | |
1.4 mg/mL | |
PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4 | |
P0C0S5, Q71UI9 | |
H2AZ1, H2az2 | |
KLH-conjugated synthetic peptide corresponding to human histone H2A.Z acetylated at lysines 4, 7, and 11. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction