Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H2BK20ac Monoclonal Antibody (GT578)
GREENER_CHOICE

Catalog No. PIMA531551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531551 Supplier Invitrogen™ Supplier No. MA531551
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Keep as concentrated solution. Positive Control: HeLa (0.4 uM Trichoststin A treatment for 18 hr). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The nucleosomes wrap further and compact DNA into chromotin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Histones play a central role in transciption regulation, DNA repair, DNA replication, and chromosomal stability. H2B has a broad antibacterial activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H2BK20ac
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone GT578
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene H2BW2
Gene Accession No. A0A2R8Y619, O60814, P06899, P0C1H6, P23527, P33778, P57053, P58876, P62807, Q16778, Q5QNW6, Q7Z2G1, Q8N257, Q93079, Q96A08, Q99877, Q99879, Q99880
Gene Alias 2610022J01Rik; Acetyl-Histone H2; Acetyl-Histone H2B K20; Acetyl-Histone H2B Lys20; AV127319; bA317E16.3; dJ160A22.3; dJ193B12.2; dJ221C16.3; dJ221C16.6; dJ221C16.8; dJ97D16.4; GL105; H2B; H2B 291A; H2B 291B; H2b 613; H2B GL105; H2B histone family member M; H2B histone family member W testis-specific; H2B histone family member W, testis specific; H2B histone family, member A; H2B histone family, member B; H2B histone family, member C; H2B histone family, member D; H2B histone family, member E; H2B histone family, member F; H2B histone family, member J; H2B histone family, member K; H2B histone family, member L; H2B histone family, member M; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member R; H2B histone family, member S; H2B histone family, member T; H2B histone family, member U, (testis-specific); H2B histone family, member W, testis-specific; H2B K; H2B type 12; H2B.1; H2B.1A; H2B.1B; H2B.2; H2B/a; H2B/b; H2B/c; H2B/d; H2B/e; H2B/f; H2B/g; H2B/h; H2B/I; H2B/j; H2B/k; H2B/l; H2B/n; H2B/q; H2B/r; H2B/S; h2B-143; h2B-221; H2b-291b; H2b-613; H2Bb; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2b-f; H2BFA; H2BFAiii; H2BFB; H2BFC; H2BFD; H2BFE; H2BFF; H2BFG; H2BFH; H2BFJ; H2BFK; H2BFL; H2BFM; H2BFN; H2BFQ; H2BFR; H2bfs; H2BFT; H2BFU; H2BFWT; H2BGL105; H2BJ; H2b-j; H2BK; H2BK12ac; H2BK15ac; H2BK5ac; H2b-l; H2BLys20ac; H2b-n; H2BQ; H2BS14; H2BS14p; HIRA-interacting protein 1; HIRA-interacting protein 2; HIRIP1; HIRIP2; HIST1H2BA; HIST1H2BB; HIST1H2BC; HIST1H2BD; Hist1h2be; Hist1h2bf; HIST1H2BG; Hist1h2bh; HIST1H2BI; Hist1h2bj; Hist1h2bk; HIST1H2BL; Hist1h2bm; Hist1h2bn; HIST1H2BO; HIST1H3I; Hist2h2be; HIST2H2BF; Hist3h2bb; histone 1, H2ba; histone 1, H2bb; histone 1, H2bc; histone 1, H2bd; histone 1, H2bg; histone 1, H2bh; histone 1, H2bi; histone 1, H2bj; histone 1, H2bk; histone 1, H2bl; histone 1, H2bm; histone 1, H2bn; histone 1, H2bo; histone 2, H2be; Histone 2B; histone 3, H2bb; histone cluster 1, H2ba; histone cluster 1, H2bb; histone cluster 1, H2bc; histone cluster 1, H2bd; histone cluster 1, H2bg; histone cluster 1, H2bh; histone cluster 1, H2bi; histone cluster 1, H2bj; histone cluster 1, H2bk; histone cluster 1, H2bl; histone cluster 1, H2bm; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 2, H2be; histone cluster 2, H2bf; histone cluster 3, H2bb; histone family member; histone H2B; histone H2B type 1-A; Histone H2B type 1-B; histone H2B type 1-C/E/F/G/I; Histone H2B type 1-C/E/G; histone H2B type 1-D; Histone H2B type 1-F/J/L; histone H2B type 1-H; Histone H2B type 1-J; histone H2B type 1-K; Histone H2B type 1-L; histone H2B type 1-M; Histone H2B type 1-N; histone H2B type 1-O; Histone H2B type 2-E; histone H2B type 2-F; Histone H2B type 3-B; histone H2B type F-M; Histone H2B type F-S; histone H2B type F-S-like; histone H2B type W-T; Histone H2B, testis; Histone H2B.1; histone H2B.1 A; Histone H2B.1 B; histone H2B.2; Histone H2B.a; histone H2B.b; Histone H2B.c; histone H2B.d; Histone H2B.e; histone H2B.f; Histone H2B.g; Histone H2B.h; Histone H2B.I; Histone H2B.j; histone H2B.k; Histone H2B.l; histone H2B.n; Histone H2B.q; histone H2B.r; Histone H2B.s; histone H2B-GL105; LOC102724334; MGC125414; MGC125415; MGC125416; MGC9388; R74621; RP1-193B12.10; STBP; T25626; testis-specific histone 2b; testis-specific histone H2B; Th2b; TSH2B; TSH2B.1
Gene Symbols H2B, H2bc1, H2BC10, H2BC11, H2BC12, H2BC13, H2BC14, H2BC15, H2BC17, H2bc18, H2BC21, H2BC3, H2BC4, H2BC5, H2BC8, H2BC9, H2BE1, H2BU1, H2BW1, H2BW2, LOC102724334
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide surrounding acetyl Lys20 of human Histone H2B. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102724334, 114483833, 128312, 158983, 255626, 286436, 3017, 3018, 440689, 8339, 8340, 8341, 8342, 8345, 8346, 8347, 8348, 8349, 85236, 8970
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.