Learn More
Invitrogen™ H2BK23ac Recombinant Rabbit Monoclonal Antibody (RM260)

Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Specifications
Specifications
| Antigen | H2BK23ac |
| Applications | ELISA, Western Blot, Luminex, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | RM260 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.09% sodium azide |
| Gene | H2BC12 |
| Gene Accession No. | A0A2R8Y619, O60814, P06899, P0C1H6, P23527, P33778, P57053, P58876, P62807, Q16778, Q5QNW6, Q7Z2G1, Q8N257, Q93079, Q96A08, Q99877, Q99879, Q99880 |
| Gene Alias | Acetyl-Histone H2B K23; Acetyl-Histone H2B Lys23; Acetyl-Histone H3 K23; Acetyl-Histone H3 Lys23; bA317E16.3; dJ160A22.3; dJ193B12.2; dJ221C16.3; dJ221C16.6; dJ221C16.8; dJ97D16.4; GL105; H2B; H2B histone family member M; H2B histone family member W testis-specific; H2B histone family member W, testis specific; H2B histone family, member A; H2B histone family, member B; H2B histone family, member C; H2B histone family, member D; H2B histone family, member E; H2B histone family, member F; H2B histone family, member J; H2B histone family, member K; H2B histone family, member L; H2B histone family, member M; H2B histone family, member N; H2B histone family, member Q; H2B histone family, member R; H2B histone family, member T; H2B histone family, member U, (testis-specific); H2B histone family, member W, testis-specific; H2B K; H2B type 12; H2B.1; H2B.1A; H2B.1B; H2B.2; H2B/a; H2B/b; H2B/c; H2B/d; H2B/e; H2B/f; H2B/g; H2B/h; H2B/j; H2B/k; H2B/l; H2B/n; H2B/q; H2B/r; H2B/S; H2Bb; H2BC10; H2BC4; H2BC6; H2BC7; H2BC8; H2BFA; H2BFAiii; H2BFB; H2BFC; H2BFD; H2BFE; H2BFF; H2BFG; H2BFH; H2BFJ; H2BFK; H2BFL; H2BFM; H2BFN; H2BFQ; H2BFR; H2BFS; H2BFT; H2BFU; H2BFWT; H2BGL105; H2BJ; H2BK; H2BLys23ac; H2BQ; HIRA-interacting protein 1; HIRA-interacting protein 2; HIRIP1; HIRIP2; HIST1H2BA; HIST1H2BB; HIST1H2BC; HIST1H2BD; HIST1H2BE; HIST1H2BF; HIST1H2BG; HIST1H2BH; HIST1H2BI; HIST1H2BJ; HIST1H2BK; HIST1H2BL; HIST1H2BM; HIST1H2BN; HIST1H2BO; HIST2H2BE; HIST2H2BF; HIST3H2BB; histone 1, H2ba; histone 1, H2bb; histone 1, H2bc; histone 1, H2bd; histone 1, H2bg; histone 1, H2bh; histone 1, H2bi; histone 1, H2bj; histone 1, H2bk; histone 1, H2bl; histone 1, H2bm; histone 1, H2bn; histone 1, H2bo; histone 2, H2be; histone 3, H2bb; histone cluster 1, H2ba; histone cluster 1, H2bb; histone cluster 1, H2bc; histone cluster 1, H2bd; histone cluster 1, H2bg; histone cluster 1, H2bh; histone cluster 1, H2bi; histone cluster 1, H2bj; histone cluster 1, H2bk; histone cluster 1, H2bl; histone cluster 1, H2bm; histone cluster 1, H2bn; histone cluster 1, H2bo; histone cluster 2, H2be; histone cluster 2, H2bf; histone cluster 3, H2bb; histone family member; histone H2B type 1-A; Histone H2B type 1-B; histone H2B type 1-C/E/F/G/I; Histone H2B type 1-D; histone H2B type 1-H; Histone H2B type 1-J; histone H2B type 1-K; Histone H2B type 1-L; histone H2B type 1-M; Histone H2B type 1-N; histone H2B type 1-O; Histone H2B type 2-E; histone H2B type 2-F; Histone H2B type 3-B; histone H2B type F-M; Histone H2B type F-S; histone H2B type F-S-like; histone H2B type W-T; Histone H2B, testis; Histone H2B.1; histone H2B.1 A; Histone H2B.1 B; histone H2B.2; Histone H2B.a; histone H2B.b; Histone H2B.c; histone H2B.d; Histone H2B.e; histone H2B.f; Histone H2B.g; Histone H2B.h; Histone H2B.j; histone H2B.k; Histone H2B.l; histone H2B.n; Histone H2B.q; histone H2B.r; Histone H2B.s; histone H2B-GL105; LOC102724334; STBP; Testis-specific histone H2B; TH2B; TSH2B; TSH2B.1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.