Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl Histone H3 (Lys27) Mouse anti-Human, Clone: 5E2.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABE670
Description
Acetyl Histone H3 (Lys27) Antibody, clone 5E2.2 is a highly specific mouse monoclonal antibody, that targets Acetyl Histone H3 (Lys27) and has been tested in Western Blotting and Immunocytochemistry and Dot Blot.
Specifications
Acetyl Histone H3 (Lys27) | |
Monoclonal | |
Please refer to lot specific datasheet. | |
HIST3H3,H3FT | |
KLH-cojugated peptide corresponding to a sequence around lysine 27 of yeast histone H3. | |
100 μg | |
Epigenetics and Nuclear Function | |
NP_009564 | |
Human | |
Purified |
Dot Blot, Immunocytochemistry, Western Blot | |
5E2.2 | |
Unconjugated | |
Mouse | |
Protein G Purified | |
RUO | |
Primary | |
Clone 5E2.2 detects histone H3 acetylated on lysine27 in human. | |
Stable for 1 year at 2-8°C from date of receipt. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction