Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ H4ac pan-acetyl (K5,K8,K12,K16) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540084
Description
H4ac pan-acetyl (K5,K8,K12,K16) Polyclonal Antibody for ICC/IF, ELISA, ARRAY
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
Specifications
| H4ac pan-acetyl (K5,K8,K12,K16) | |
| Polyclonal | |
| Unconjugated | |
| H4C1 | |
| acetyl Histone H4; Acetyl-H4 K5 K8 K12 K16; Acetyl-H4 Lys5 Lys8 Lys12 Lys16; Acetyl-Histone H4 K5 K8 K12 K16; Acetyl-Histone H4 Lys5 Lys8 Lys12 Lys16; FO108; H4; H4 ac pan-acetyl; H4 histone family, member A; H4 histone family, member I; H4 histone family, member J; H4 histone family, member M; H4 histone family, member N; H4 histone, family 2; H4/A; H4/B; H4/C; H4/D; H4/E; H4/G; H4/H; H4/I; H4/J; H4/K; H4/M; H4/n; H4/O; H4/p; H4-12; H4-16; H4-53; H4ac; H4c1; H4c11; H4C12; H4C13; H4c14; H4C15; H4c2; H4c3; H4c4; H4C5; H4c6; H4C8; H4c9; H4f16; H4F2; H4FA; H4FB; H4FC; H4FD; H4FE; H4FG; H4FH; H4FI; H4FJ; H4FK; H4FM; H4FN; H4FO; H4K12; H4K12a; H4K12ac; H4K16; H4K16ac; H4K5; H4K5,8,12,16ac; H4K5ac; H4K8; H4K8ac; H4Lys5,8,12,16ac; H4M; HIST1H4; HIST1H4A; Hist1h4b; HIST1H4C; Hist1h4d; HIST1H4E; Hist1h4f; Hist1h4h; Hist1h4i; HIST1H4J; Hist1h4k; HIST1H4L; Hist1h4m; Hist2h4; HIST2H4A; HIST2H4B; Hist4h4; histone 1, H4a; histone 1, H4b; histone 1, H4e; histone 1, H4i; histone 2, H4; histone 2, H4a; Histone 4; Histone 4 family, member M; histone 4, H4; histone cluster 1, H4a; histone cluster 1, H4b; histone cluster 1, H4e; histone cluster 1, H4i; histone cluster 2, H4; histone cluster 2, H4a; Histone Cluster 4; histone cluster 4, H4; histone family member; histone H4; Histone H4 acetyl; Histone H4A; Histone H4K12; Histone H4K16; Histone H4K5; Histone H4K8; histone IV, family 2; X04652 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 121504, 319158, 326619, 326620, 8294, 8359, 8363, 8366, 8367, 8370, 97122 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Peptide Array, ELISA, Immunocytochemistry | |
| 0.76 mg/mL | |
| PBS with 0.05% ProClin 300, 0.05% sodium azide; pH 7.4 | |
| P62805, P62806 | |
| H4-16, H4C1, H4C11, H4C14, H4C2, H4C5, H4C9 | |
| KLH-conjugated synthetic peptide corresponding to human the region of histone H4 containing the acetylated lysines 5, 8, 12 and 16 (H4K5,8,12,16ac). | |
| 50 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction