Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl-Lysine Antibody (1C6), Novus Biologicals™
SDP

Catalog No. NB10074339 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074339 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074339 Supplier Novus Biologicals Supplier No. NB10074339
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Acetyl-Lysine Monoclonal antibody specifically detects Acetyl-Lysine in Nonspecies specific samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Dot Blot, ChIP assay.

Specifications

Antigen Acetyl-Lysine
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Dot Blot
Classification Monoclonal
Clone 1C6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 3 ug, Dot Blot 1:100 - 1:2000, Chromatin Immunoprecipitation (ChIP) 10 ug
Gene Alias acetyl Lysine, AC-LYS-OH, Lysine, N-acetyl lysine
Host Species Mouse
Immunogen Synthetic peptide sequence surrounding the acetylated lysine 9 of histone H3.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Acetylated Lysine (1C6)
Target Species Non-species specific, Mouse, Non-species specific, Monkey
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.