Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetyl-Lysine Antibody (7F8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB11096876H
Description
Acetyl-Lysine Monoclonal antibody specifically detects Acetyl-Lysine in Nonspecies specific samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Acetyl-Lysine | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
Mouse | |
Protein G purified | |
Primary | |
Non-species specific | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
7F8 | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
acetyl Lysine, AC-LYS-OH, Lysine, N-acetyl lysine | |
Acetylated KLH | |
0.1 mL | |
Detects proteins containing acetylated lysine residues in ELISA and western blots. Does not detect non-acetylated lysine residues. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction