Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetyl MLL1 (KMT2A) (Lys 1130/1133) Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABE34425UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABE344-25UG 25 μg
ABE344-100UG 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABE344-25UG Supplier MilliporeSigma Supplier No. ABE34425UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Acetyl MLL1 (KMT2A) (Lys 1130/1133), Cat. No. ABE344, is a highly specific rabbit polyclonal antibody that targets Histone-lysine N-methyltransferase 2A (MLL1) acetylated on Lysine 1130/1133 and has been tested for use in Dot Blot, Peptide Inhibition Assay, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetyl MLL1 (KMT2A) (Lys 1130/1133)
Applications Dot Blot, Inhibition Assays, Western Blot
Classification Polyclonal
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.09% sodium azide.
Gene Alias Histone-lysine N-methyltransferase 2A;EC: 2.1.1.43;Lysine N-methyltransferase 2A;ALL-1;CXXC-type zinc finger protein 7;Myeloid/lymphoid or mixed-lineage leukemia;Myeloid/lymphoid or mixed-lineage leukemia protein 1;Trithorax-like protein;Zinc finger protein HRX
Gene Symbols KMT2A;ALL1;CXXC7;HRX;HTRX;MLL;MLL1;TRX1
Host Species Rabbit
Immunogen KLH-cpnugated linear peptide corresponding to 12 amino acids surrounding acetylated lysines 1130 and 1133 from the N-terminal half of human Histone-lysine N-methyltransferase 2A.
Molecular Weight of Antigen 431.76 kDa calculated.
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005924
Test Specificity This rabbit polyclonal antibody specifically detects human Histone-lysine N-methyltransferase 2A acetylated on lysine 1130 and 1133.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.