Learn More
Invitrogen™ Acetyl-SMAD2/SMAD3 (Lys19) Polyclonal Antibody

Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is >95% (by SDS-PAGE).
Specifications
Specifications
| Antigen | Acetyl-SMAD2/SMAD3 (Lys19) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | SMAD2 |
| Gene Accession No. | O70436, P84022, P84025, Q15796, Q62432, Q8BUN5 |
| Gene Alias | 7120426M23Rik; AU022421; hMAD-2; hMAD-3; hSMAD2; hSMAD3; HSPC193; HsT17436; JV15-2; JV18; JV18-1; LDS1C; LDS3; MAD (mothers against decapentaplegic, Drosophila) homolog 3; MAD homolog 2; MAD homolog 3; mad homolog JV15-2; mad protein homolog; MAD, mothers against decapentaplegic homolog 3; mad3; MADH2; MADH3; MADR2; mad-related protein 2; mMad2; mMad3; mother against DPP homolog 2; mothers against decapentaplegic homolog 2; Mothers against decapentaplegic homolog 3; mothers against DPP homolog 2; mothers against DPP homolog 3; pSMAD2; Sma- and Mad-related protein 2; SMA- and MAD-related protein 3; SMAD; Smad 2; Smad 3; SMAD family member 2; SMAD family member 3; SMAD, mothers against DPP homolog 2; SMAD, mothers against DPP homolog 3; Smad2; Smad-2; Smad3 |
| Gene Symbols | SMAD2, SMAD3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.