Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ acetyl SMC3 (Lys105/106), Mouse, Unlabeled, Clone: 21A7,

Catalog No. MABE1073 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE1073 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1073 Supplier MilliporeSigma™ Supplier No. MABE1073
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Structural maintenance of chromosomes protein 3 (UniProt Q9UQE7; also known as Bamacan, Basement membrane-associated chondroitin proteoglycan, Chondroitin sulfate proteoglycan 6, Chromosome-associated polypeptide, hCAP, SMC protein 3, SMC-3) is encoded by the SMC3 (also known as BAM, BMH, CDLS3, CSPG6, SMC3L1) gene (Gene ID 9126) in human. Cohesion is a nuclear tripartite complex composed of Rad21 and two structural maintenance of chromosomes proteins, SMC1 and SMC3. Cohesin plays an important role in sister chromatid cohesion, as well as in DNA repair and gene expression regulation. To establish sister chromatid cohesion during S phase, SMC3 is acetylated (on K105 and K106 in human) by two cohesin acetyltransferases (CoATs), ESCO1 and ESCO2, that differ in their N-terminal domains and expression during development and across the cell cycle. ChIP-seq analysis reveals that ESCO1 is recruited by cohesin to over 11,000 chromatin sites occupied by cohesin and CTCF that mediate long-range chromatin interactions and regulate transcription globally. On ther other hand, ESCO2 is infrequently enriched at sites targeted by REST/NRSF (RE1-silencing transcription factor/neuron-restrictive silencer factor) that represses transcription of neuron-specific genes. SMC3 acetylation is thought to induce conformational changes of the dimeric hinge structure during S phase and to allow dimer opening and loading of DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen acetyl SMC3 (Lys105/106)
Applications ChIP Assay, ELISA, Western Blot
Classification Monoclonal
Clone 21A7
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2b in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene SMC3, BAM, BMH, CDLS3, CSPG6, SMC3L1
Gene Accession No. Q9UQE7
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the target region sequence of human/mouse/rat SMC3 containing acetylated Lys105 and Lys106.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_005436
Test Specificity Clone 21A7 bound Lys105/106-diacetylated SMC3 peptide with 2-fold higher affinity than Lys105-monoacetylated SMC3 peptide (Kd = 0.04 and 0.09μg/mL, respectively), while displaying much reduced affinity toward Lys105-monoacetylated (Kd = 0.73μg/mL) and little affinity toward nonacetylated (Kd = 2.99μg/mL) peptide (Rahman, S., et al. (2015). Proc. Natl. Acad. Sci. U. S. A. 112(36):11270-11275).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.