Learn More
Acetyl-SMC3 (Lys105, Lys106) Monoclonal Antibody, Active Motif

Mouse Monoclonal Antibody
$156.65 - $620.58
Specifications
Antigen | Acetyl-SMC3 (Lys105, Lys106) |
---|---|
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-255-6793
![]() ![]() |
Active Motif
65729 |
10 μg |
Each for $156.65
|
|
|||||
50-255-6794
![]() ![]() |
Active Motif
65728 |
100 μg |
Each for $620.58
|
|
|||||
Description
Some products may be shipped at room temperature. This will not affect their stability or performance. Avoid repeated freeze/thaw cycles by aliquoting items into single-use fractions for storage at -20°C for up to 2 years. Keep all reagents on ice when not in storage.
Acetyl-SMC3 (Lys105, Lys106) Monoclonal antibody specifically detects Acetyl-SMC3 (Lys105, Lys106) in Human, Mouse samples. It is validated for Western BlotSpecifications
Acetyl-SMC3 (Lys105, Lys106) | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide | |
Bam; Bamacan; basement membrane chondroitin sulfate proteoglycan; basement membrane-associated chondroitin proteoglycan; BMH; CDLS3; chondroitin sulfate proteoglycan 6; chondroitin sulfate proteoglycan 6 (bamacan); chromosome segregation protein SmcD; Chromosome-associated polypeptide; cspg6; HCAP; im:7142991; mad member-interacting protein 1; Mmip1; SMC protein 3; Smc3; SMC-3; SMC3 protein; SMC3L1; SmcD; structural maintenace of chromosomes 3; structural maintenance of chromosomes 3; structural maintenance of chromosomes protein 3; structural maintenance of chromosomes protein 3-like protein; wu:fb22e01; wu:fc30d07 | |
This SMC3 antibody was raised against a peptide in the C-terminal region of human SMC3. | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
SMC3 |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q9CW03, Q9UQE7 | |
13006, 9126 | |
IgG1 | |
Protein G | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.