Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylacetonatobis(ethylene)rhodium(I), 99%

Catalog No. ac363381000
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle

Catalog No. AC363381000

Supplier: Thermo Scientific Chemicals 363381000

Only null left
Add to Cart

CAS: 12082-47-2 | C9H15O2Rh | 258.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12082-47-2
Molecular Formula C9H15O2Rh
Molecular Weight (g/mol) 258.12
MDL Number MFCD00015354
InChI Key FLRBEQQDEGBCJS-FGSKAQBVSA-M
Synonym acetylacetonatobis ethylene rhodium i, bis ethylene 2,4-pentanedionato rhodium i, acetylacetonatobis ethylene rhodium, ethylene; z-4-oxopent-2-en-2-olate; rhodium
PubChem CID 11630270
IUPAC Name ethene;(Z)-4-oxopent-2-en-2-olate;rhodium
SMILES [Rh+].C=C.C=C.C\C([O-])=C\C(C)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 141.0°C
Color Orange
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 100 mg
Formula Weight 258.13
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Acetylacetonatobis(ethylene)rhodium(I)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
EINECSNumber 235-147-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.