Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylcholine Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200271 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017625 0.1 mL
NB10064656 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017625 Supplier Novus Biologicals Supplier No. NB100646560.1ML
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acetylcholine Polyclonal specifically detects Acetylcholine in All Species samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen Acetylcholine
Applications Immunohistochemistry (Frozen), ELISA, Immunohistochemistry
Classification Polyclonal
Concentration 5.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1:50-1:500, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Alias [2 (acetyloxy)ethyl]trimethylammonium, ACh
Host Species Rabbit
Immunogen Synthetic acetylcholine conjugated to Keyhole limpet Haemocyanin.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Rabbit anti acetylcholine polyclonal antibody recognizes the small cationic neurotransmitter acetylcholine, involved in the transmission of nerve impulses across synapses. The structure of acetylcholine is preserved across the animal kingdom and the Rabbit anti acetylcholine is expected to recognize acetylcholine from all species posessing this neurotransmitter.
Target Species All species
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.