Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetylcholinesterase/ACHE Antibody, Alexa Fluor™ 750, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB600911AF750
Description
Acetylcholinesterase/ACHE Polyclonal specifically detects Acetylcholinesterase/ACHE in Bovine samples. It is validated for Western Blot, ELISA.Specifications
Acetylcholinesterase/ACHE | |
Polyclonal | |
Western Blot, ELISA | |
acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group | |
Acetylcholinesterase/ACHE [Bovine Erythrocytes] (Uniprot: P23795) | |
0.1 mL | |
Cellular Markers, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
43 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Goat | |
Multi-step | |
RUO | |
Primary | |
Bovine | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction