Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acetylcholinesterase/ACHE Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB1001519
Description
Acetylcholinesterase/ACHE Polyclonal specifically detects Acetylcholinesterase/ACHE in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Peptide ELISA, Immunohistochemistry Free-Floating.Specifications
Acetylcholinesterase/ACHE | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
acetylcholinesterase, acetylcholinesterase (Yt blood group), AChE, apoptosis-related acetylcholinesterase, ARACHE, EC 3.1.1, EC 3.1.1.7, N-ACHE, YT, Yt blood group | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
This antibody is expected to recognise isoform NP_000656.1 only (the ubiquitously expressed, hydrophillic form). | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, ELISA | |
0.5 mg/mL | |
Western Blot 0.3 - 1 μg/mL, Flow Cytometry 10 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Peptide ELISA Detection limit 1:32000, Immunohistochemistry Free-Floating | |
NP_000656. | |
ACHE | |
Peptide with sequence QFDHYSKQDRCSDL corresponding to C-Terminus according to NP_000656.1. | |
0.1 mg | |
Cellular Markers | |
43 | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction