Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acetylcholinesterase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA515010
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA515010 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA515010 Supplier Invitrogen™ Supplier No. PA515010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Acetylcholinesterase Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

Acetylcholinesterase hydrolyzes the neurotransmitter, acetylcholine at neuromuscular junctions and brain cholinergic synapses, and thus terminates signal transmission. The Protein is also found on the red blood cell membranes, where it constitutes the Yt blood group antigen. Acetylcholinesterase exists in multiple molecular forms which possess similar catalytic properties, but differ in their oligomeric assembly and mode of cell attachment to the cell surface. The major form of acetylcholinesterase found in brain, muscle and other tissues is the hydrophilic species, which forms disulfide-linked oligomers with collagenous, or lipid-containing structural subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetylcholinesterase
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ACHE
Gene Accession No. P22303
Gene Alias ACE-1; ACEE; Acetylcholinesterase; acetylcholinesterase (Cartwright blood group); acetylcholinesterase (Yt blood group); acetylcholinesterase T-subunit; ACHE; ache protein; apoptosis-related acetylcholinesterase; ARACHE; EC 3.1.1.7; glycolipid-anchored form of acetylcholinesterase; N-ACHE; OTTHUMP00000211349; OTTHUMP00000211356; YT; Yt blood group; zgc:92550; ziehharmonika; zim
Gene Symbols ACHE
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 147-175 amino acids from the N-terminal region of human ACHE.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.