Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Acetylcholinesterase Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518348
Description
This antibody is predicted to react with bovine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Acetylcholinesterase hydrolyzes the neurotransmitter, acetylcholine at neuromuscular junctions and brain cholinergic synapses, and thus terminates signal transmission. The Protein is also found on the red blood cell membranes, where it constitutes the Yt blood group antigen. Acetylcholinesterase exists in multiple molecular forms which possess similar catalytic properties, but differ in their oligomeric assembly and mode of cell attachment to the cell surface. The major form of acetylcholinesterase found in brain, muscle and other tissues is the hydrophilic species, which forms disulfide-linked oligomers with collagenous, or lipid-containing structural subunits.
Specifications
| Acetylcholinesterase | |
| Polyclonal | |
| Unconjugated | |
| ACHE | |
| ACE-1; ACEE; Acetylcholinesterase; acetylcholinesterase (Cartwright blood group); acetylcholinesterase (Yt blood group); acetylcholinesterase T-subunit; ACHE; ache protein; apoptosis-related acetylcholinesterase; ARACHE; EC 3.1.1.7; glycolipid-anchored form of acetylcholinesterase; N-ACHE; OTTHUMP00000211349; OTTHUMP00000211356; YT; Yt blood group; zgc:92550; ziehharmonika; zim | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 43 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P22303 | |
| ACHE | |
| Synthetic peptide sequence (QFDHYSKQDRCSDL) corresponding to the C-terminus amino acids of ACHE (aa 601-614). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction