Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acetylcholinesterase Recombinant Rabbit Monoclonal Antibody (HL1102)
GREENER_CHOICE

Catalog No. PIMA542348
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542348 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542348 Supplier Invitrogen™ Supplier No. MA542348
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Acetylcholinesterase hydrolyzes the neurotransmitter, acetylcholine at neuromuscular junctions and brain cholinergic synapses, and thus terminates signal transmission. The Protein is also found on the red blood cell membranes, where it constitutes the Yt blood group antigen. Acetylcholinesterase exists in multiple molecular forms which possess similar catalytic properties, but differ in their oligomeric assembly and mode of cell attachment to the cell surface. The major form of acetylcholinesterase found in brain, muscle and other tissues is the hydrophilic species, which forms disulfide-linked oligomers with collagenous, or lipid-containing structural subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acetylcholinesterase
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1102
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ACHE
Gene Accession No. P21836, P22303, P37136
Gene Alias ACE-1; ACEE; Acetylcholinesterase; acetylcholinesterase (Cartwright blood group); acetylcholinesterase (Yt blood group); acetylcholinesterase T-subunit; ACHE; ache protein; apoptosis-related acetylcholinesterase; ARACHE; EC 3.1.1.7; glycolipid-anchored form of acetylcholinesterase; N-ACHE; OTTHUMP00000211349; OTTHUMP00000211356; YT; Yt blood group; zgc:92550; ziehharmonika; zim
Gene Symbols ACHE
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human AChE. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11423, 43, 83817
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.