Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylene-PEG4-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals

Catalog No. p-7042994
Click to view available options
Quantity:
5 mg
25 mg

A carboxyrhodamine 110 based labeling reagent

A carboxyrhodamine 110 based labeling reagent; reacts with azides via a copper-catalyzed click reactions. Carboxyrhodamine 110 is a bright and photostable green fluorescent label that can be excited at a wavelength of 488nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1615739-95-1
Molecular Formula C32H33N3O8
MDL Number MFCD22380741
Synonym Acetylene-Fluor 488; Rhodamine 110-PEG4-alkyne

Specifications

Color Red
Quantity 5 mg
Solubility Information Soluble in DMSO,DMF,or methanol
Formula Weight 587.63
Physical Form Solid
Chemical Name or Material Acetylene-PEG4-carboxyrhodamine 110 conjugate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.