Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylene-PEG4-maleimide

Catalog No. AAJ64859MB
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64859MB 25 mg
AAJ64859MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ64859MB Supplier Thermo Scientific Chemicals Supplier No. J64859MB
Add to Cart

CAS: 1609651-90-2 | C18H26N2O7 | 382.41 g/mol

Heterobifunctional cross linker enabling the connection of an azide containing compound or biomolecule to one with a thiol. The maleimide group will react preferentially with thiols to form a covalent adduct and the terminal alkyne will react with azides via copper-catalyzed 1,3-dipolar cycloaddition click chemistry. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification cysteine containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character. A bifunctional linker with sulfhydryl reactive and azide reactive groups. Contains a PEG spacer to increase solubility

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Heterobifunctional cross linker enabling the connection of an azide containing compound or biomolecule to one with a thiol. The maleimide group will react preferentially with thiols to form a covalent adduct and the terminal alkyne will react with azides via copper-catalyzed 1,3-dipolar cycloaddition click chemistry. The hydrophilic PEG spacer adds to the water solubility of this reagent, allowing for easy modification cysteine containing biomolecules such as antibodies, proteins and peptides with the terminal alkyne. The PEG spacer also reduces aggregation of modified proteins stored in solution by adding hydrophilic character. A bifunctional linker with sulfhydryl reactive and azide reactive groups. Contains a PEG spacer to increase solubility

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1609651-90-2
Molecular Formula C18H26N2O7
Molecular Weight (g/mol) 382.41
MDL Number MFCD22380740
InChI Key GHEBECZRHJXPTL-UHFFFAOYSA-N
Synonym acetylene-peg4-maleimide, propargyl-peg4-maleimide, alkyne-peg4-maleimide hplc
PubChem CID 91757777

Specifications

Color Colorless to Yellow
Quantity 25 mg
Solubility Information Soluble in water.
Formula Weight 382.41
Physical Form Viscous Liquid or Solid
Chemical Name or Material Acetylene-PEG4-maleimide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.