Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylferrocene, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
1271-55-2
C12H12FeO
228.07
MFCD00001432
SPKJCVZOZISLEI-UHFFFAOYSA-N
1271-55-2
C12H12FeO
228.07
MFCD00001432
SPKJCVZOZISLEI-UHFFFAOYSA-N
Boiling Point 160.0°C to 163.0°C (4.0hPa)
Quantity 25g
Infrared Spectrum Authentic
Color Orange
Packaging Glass bottle
Melting Point 81.0°C to 86.0°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 228.07
Physical Form Needle-Like Crystalline Powder
Percent Purity 95%
Chemical Name or Material Acetylferrocene
Show More Show Less

GHS H Statement:
Toxic in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger