Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylferrocene, 95%, Thermo Scientific™

Catalog No. AC147991000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC147991000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC147991000 Supplier Thermo Scientific Chemicals Supplier No. 147991000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1271-55-2
Molecular Formula C12H12FeO
Molecular Weight (g/mol) 228.07
MDL Number MFCD00001432
InChI Key SPKJCVZOZISLEI-UHFFFAOYSA-N
IUPAC Name 1-Acetylferrocene
SMILES [Fe].c1cccc1.CC(=O)c1cccc1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 81.0°C to 86.0°C
Color Orange
Boiling Point 160.0°C to 163.0°C (4.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 228.07
Percent Purity 95%
Physical Form Needle-like Crystalline Powder
Chemical Name or Material Acetylferrocene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.