Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylsalicylsalicylic acid, 97%

Catalog No. p-49886
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle

CAS: 530-75-6 | C16H12O6 | 300.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-75-6
Molecular Formula C16H12O6
Molecular Weight (g/mol) 300.26
MDL Number MFCD00143537
InChI Key DDSFKIFGAPZBSR-UHFFFAOYSA-N
Synonym 2-2-acetoxybenzoyl oxy benzoic acid, acetylsalicylsalicylic acid, acesalum, acetylsalicylsalicyic acid, diplosal acetate, unii-vbe72mcp5l, 2-carboxyphenyl 2-acetyloxy benzoate, diplosalsalate, ccris 1740, benzoic acid, 2-acetyloxy-, 2-carboxyphenyl ester
PubChem CID 10745
IUPAC Name 2-(2-acetyloxybenzoyl)oxybenzoic acid
SMILES CC(=O)OC1=CC=CC=C1C(=O)OC2=CC=CC=C2C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 145°C to 160°C
Assay Percent Range 96% min. (As is) Calculated (HPLC)
Packaging Glass bottle
Quantity 250 mg
Beilstein 10,IV,165
Merck Index 14,1
Solubility Information Solubility in water: .. Other solubilities: soluble in alkaline solutions,soluble in alcohol,practically insoluble in benzene
Formula Weight 300.26
Percent Purity 97%
Chemical Name or Material Acetylsalicylsalicylic acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
EINECSNumber 208-493-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.