Learn More
ACF1/BAZ1A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301318A

Description
The recommended shelf life for this product is 1 year from date of receipt.
BAZ1A may play a role in transcriptional regulation. May be involved in the formation or maintece of heterochromatin playing a critical role in developmental control.Specifications
ACF1/BAZ1A | |
Polyclonal | |
Unconjugated | |
BAZ1A | |
ACF1, ATP-dependent chromatin remodeling protein, ATP-dependent chromatin-remodeling protein, ATP-utilizing chromatin assembly and remodeling factor 1, bromodomain adjacent to zinc finger domain, 1A, CHRAC subunit ACF1, hACF1, hWALp1, WALp1, WCRF180, williams syndrome transcription factor-related chromatin-remodeling factor 180 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11177 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9NRL2 | |
BAZ1A | |
Between 725 and 775 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.