Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Achaete scute homolog 3 (ASCL3) Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
SDP

Catalog No. 502543913
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3913 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3913 Supplier OriGene Supplier No. CF811864
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Achaete scute homolog 3 (ASCL3) Monoclonal antibody specifically detects Achaete scute homolog 3 (ASCL3) in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Achaete scute homolog 3 (ASCL3)
Applications Western Blot
Classification Monoclonal
Clone OTI1C5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene ITM2A
Gene Accession No. O43736, Q61500
Gene Alias achaete-scute complex homolog 3; achaete-scute complex homolog 3 (Drosophila); achaete-scute complex homolog-like 3; achaete-scute family bHLH transcription factor 3; achaete-scute homolog 3; Ascl3; ASH-3; ASH3A; bHLH transcription factor Sgn-1 (Salivary Glands 1); bHLH transcriptional regulator Sgn-1; bHLHa42; class A basic helix-loop-helix protein 42; HASH3; ICRFP703B1614Q5.4; ICRFP703N2430Q5.4; mASH3; mASH-3; Sgn1
Host Species Mouse
Immunogen Full length human recombinant protein of human ITM2A (NP_004858) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16431, 317218, 9452
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.