Learn More
Acid Alcohol, Orth's Decolorizing Fluid, 3% HCl in 70% Alcohol, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Physical Form | Solution |
| CAS Min % | 26.0, 66.0, 3.0, 2.9 |
| CAS Max % | 28.0, 67.0, 4.0, 3.1 |
| Packaging | Natural Poly Bottle |
| Color | Colorless |
| pH | Acidic |
| Quantity | 500 mL |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 3% HCl |
| Chemical Name or Material | Acid Alcohol, Orth's Decolorizing Fluid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.