Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Acid and Base Neutralizing Polymers
Click to view available options
Type:
Acid Neutralizing Polymer
Base Neutralizing Polymer
Color:
Turns orange
Turns purple
Description
- Provides safe spill control
- Polymers neutralize and change color when neutralization has taken place
- Acid neutralizer turns purple
- Base neutralizer turns orange
- Non-toxic, non-corrosive and non-hazardous
Specifications
Specifications
Color | Turns purple |
Capacity (English) | 2.6 qt. |
Type | Acid Neutralizing Polymer |
For Use With (Application) | Acid Chemical Spills |
Packaging Quantity | 1 Ea. |
Dimensions (L x W) | 12.5 x 9.5 in. |
Length (English) | 12.5 in. |
Width (English) | 12.5 in. |
Height (English) | 9.5 in |
Depth (English) | 12.5 in |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction