Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acid Blank, 5% (v/v) Hydrochloric Acid, Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
4 L
10 L
Specifications
Specifications
| Chemical Name or Material | Acid Blank, Hydrochloric Acid |
| Packaging | Cubitainer |
| CAS | 7647-01-0, 7732-18-5 |
| Concentration or Composition Notes | 5% (v/v) |
| CAS Min % | 94.0, 4.0 |
| CAS Max % | 96.0, 6.0 |
| Physical Form | Liquid |
| Color | Colorless |
| Melting Point | Approximately 0°C |
| Quantity | 4 L |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction