Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Digestion Reagent (with Mercury), For Organic Nitrogen, LabChem™

Catalog No. p-5388347
Click to view available options
Quantity:
1 L
4 L

Specifications

Chemical Name or Material Acid Digestion Reagent (with Mercury)
Color Colorless
Name Note For Organic Nitrogen
CAS Max % 56.9, 29.5, 13.4, 0.2
CAS 7664-93-9, 7778-80-5, 7783-35-9
Linear Formula HgSO4
UN Number UN2796
Density >1.2g/mL
Solubility Information Soluble in water
Formula Weight 296.65
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.2g/mL
Decomposition Information Sulfur compounds; Mercury; Thermal decomposition generates: Corrosive vapors
Quantity 1 L
Physical Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.