Learn More
StatLab Pneumocystis Control Slides
Click to view available options
:
Pneumocystic Controls
Description
- Standardize and satisfy regulatory requirements with Pneumocystis Stain Control Slides
- These control slides simulate clinical specimens of homogenate of lung tissue
- Scan quickly with high organism counts
- Not for use with monoclonal antibody fluorescent assays
- 10 slides/box
Specifications
Specifications
Quantity | 10 Slides/Box |
Type | Control Slide |