Learn More
Dentec Safety™ Acid Gas and Organic Vapor Cartridge
Dentec Safety™ Acid Gas and Organic Vapor Cartridge features unique space plenums that allow users to access the entire filter material reducing breathing resistance, providing longer cartrdige life, reducing costs and improving productivity
$56.50 - $796.00
Specifications
Material | Plastic |
---|---|
Product Type | Respirator Accessories |
For Use With (Equipment) | Masks |
Includes | Cartridges and filters |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
19-222-513
![]() |
Dentec Safety™
158T26 |
6/Box |
Pack of 6 for $56.50
|
|
|||||
19-222-514
![]() |
Dentec Safety™
158T26B100 |
100/Box |
Case of 100 for $796.00
|
|
|||||
Description
- Offset connector allows the user to rotate the cartridge into 3 different positions improves visibility and allows face shields or welding helmets to be worn without interference
- Saf-Twist design quickly attaches the cartridge to any Dentec Safety half-mask or full face air-purifying respirator Facepiece.
Specifications
Plastic | |
Respirator Accessories | |
Masks | |
Cartridges and filters |