Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acid Phosphatase 1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582558
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582558 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582558 Supplier Invitrogen™ Supplier No. PA582558
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ENWRVDSAAT SGYEIGNPPD YRGQSCMKRH GIPMSHVARQ ITKEDFATFD YILCMDESNL RDLNRKSNQV KTCKAKIELL GSYDPQKQLI IEDPYYGNDS DFETVYQ Highest antigen sequence indentity to the following orthologs: Mouse - 89%, Rat - 89%.

Low molecular weight protein tyrosine phosphtases, including the human red cell phosphatase, HCPTP, are widely expressed cytosolic proteins of approximately 18 kDa that exist in distinct isoforms and are highly selective for phosphotyrosine over phosphoserine or phosphotyrosine. The HCPTP-A and HCPTP-B proteins expressed in human red cells and placenta are fast and slow forms of red cell acid phosphatase5. Like other tyrosine phosphatases, HCPTP is sensitive to inhibition by vanadate, and has a catalytic mechanism that involves formation of a cystinylphosphate intermediate6. The crystal structure of the bovine heart derived enzyme BHPTP shows a four-stranded central parallel b sheet with flanking a helices and an active site cysteine residue in the typical tyrosine phosphatase sequence context, CXXXXXR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acid Phosphatase 1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ACP1
Gene Accession No. P24666, P41498, Q9D358
Gene Alias 4632432E04Rik; acid phosphatase; acid phosphatase 1; acid phosphatase 1, soluble; acid phosphatase of erythrocyte; acp1; Acp-1; Acp1-PA; adipocyte acid phosphatase; Adult cuticle protein 1; AI427468; CG7216; CG7216-PA; cytoplasmic phosphotyrosyl protein phosphatase; Dacp-1; Dmel\CG7216; Dmel_CG7216; HAAP; HAAP betaÊ; hypothetical protein; LMW-PTP; LMW-PTPase; LMW-PTP-I low molecular weight phosphotyrosine protein phosphatase isozyme 1; Low molecular weight cytosolic acid phosphatase; low molecular weight phosphotyrosine protein phosphatase; MGC111030; MGC3499; phosphatase 1, soluble; protein tyrosine phosphatase; ptpase; RCJMB04_11a4; Red cell acid phosphatase 1; RH03221p; testicular secretory protein Li 37; Unknown (protein for MGC:128334)
Gene Symbols ACP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ACP1. Recombinant protein control fragment (Product #RP-91565).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11431, 24161, 52
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.