Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Acid Phosphatase 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595936
Description
Immunogen sequence: GNICRSPIAE AVFRKLVTDQ NISENWRVDS AATSGYEIGN PPDYRGQSCM KRHGIPMSHV ARQITKEDFA TFDYILCMDE SNLRDLNRKS NQVKTCKAKI ELLGSYDPQK QLIIEDPYYG NDSDFETVYQ; Positive Samples: Mouse brain, Mouse liver, Mouse skin; Cellular Location: Cytoplasm.
Low molecular weight protein tyrosine phosphtases, including the human red cell phosphatase, HCPTP, are widely expressed cytosolic proteins of approximately 18 kDa that exist in distinct isoforms and are highly selective for phosphotyrosine over phosphoserine or phosphotyrosine. The HCPTP-A and HCPTP-B proteins expressed in human red cells and placenta are fast and slow forms of red cell acid phosphatase5. Like other tyrosine phosphatases, HCPTP is sensitive to inhibition by vanadate, and has a catalytic mechanism that involves formation of a cystinylphosphate intermediate6. The crystal structure of the bovine heart derived enzyme BHPTP shows a four-stranded central parallel b sheet with flanking a helices and an active site cysteine residue in the typical tyrosine phosphatase sequence context, CXXXXXR.
Specifications
Acid Phosphatase 1 | |
Polyclonal | |
Unconjugated | |
ACP1 | |
4632432E04Rik; acid phosphatase; acid phosphatase 1; acid phosphatase 1, soluble; acid phosphatase of erythrocyte; acp1; Acp-1; Acp1-PA; adipocyte acid phosphatase; Adult cuticle protein 1; AI427468; CG7216; CG7216-PA; cytoplasmic phosphotyrosyl protein phosphatase; Dacp-1; Dmel\CG7216; Dmel_CG7216; HAAP; HAAP betaÊ; hypothetical protein; LMW-PTP; LMW-PTPase; LMW-PTP-I low molecular weight phosphotyrosine protein phosphatase isozyme 1; Low molecular weight cytosolic acid phosphatase; low molecular weight phosphotyrosine protein phosphatase; MGC111030; MGC3499; phosphatase 1, soluble; protein tyrosine phosphatase; ptpase; RCJMB04_11a4; Red cell acid phosphatase 1; RH03221p; testicular secretory protein Li 37; Unknown (protein for MGC:128334) | |
Rabbit | |
Affinity chromatography | |
RUO | |
11431, 52 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P24666, Q9D358 | |
ACP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 15-144 of human ACP1 (NP_0042911). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction