Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acid Phosphatase 2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $404.50
Specifications
Antigen | Acid Phosphatase 2 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145477
|
Invitrogen™
PA5145477 |
200 μL |
Each for $404.50
|
|
|||||
FEPA5145478
|
Invitrogen™
PA5145478 |
20 μL |
Each for $191.80
|
|
|||||
Description
This gene encodes the beta subunit of lysosomal acid phosphatase (LAP). LAP is chemically and genetically distinct from red cell acid phosphatase. The encoded protein belongs to a family of distinct isoenzymes which hydrolyze orthophosphoric monoesters to alcohol and phosphate. Mutations in this gene or in the related alpha subunit gene cause acid phosphatase deficiency. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene.Specifications
Acid Phosphatase 2 | |
Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P11117, P20611, P24638 | |
11432, 24162, 53 | |
Recombinant Human ACP2 protein, Tyr40-Ala250 (Accession #P11117). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
acid phosphatase 2, lysosomal; Acid phosphatase 2, lysozymal; Acp2; Acp-2; LAP; LMW-PTP-II low molecular weight phosphotyrosine protein phosphatase isozyme 2; lysosomal acid phosphatase; Unknown (protein for MGC:142461) | |
Acp2 | |
IgG | |
Protein A | |
Acp2 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title