Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acid Red 114, TCI America™
SDP

Catalog No. A187425G
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 6459-94-5
Molecular Formula C37H28N4Na2O10S3
Molecular Weight (g/mol) 830.809
MDL Number MFCD00003894
InChI Key PVLKQJLDEXCKNY-UHFFFAOYSA-L
PubChem CID 5479172
IUPAC Name disodium;8-[[2-methyl-4-[3-methyl-4-[[4-(4-methylphenyl)sulfonyloxyphenyl]diazenyl]phenyl]phenyl]hydrazinylidene]-7-oxonaphthalene-1,3-disulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OC2=CC=C(C=C2)N=NC3=C(C=C(C=C3)C4=CC(=C(C=C4)NN=C5C(=O)C=CC6=CC(=CC(=C65)S(=O)(=O)[O-])S(=O)(=O)[O-])C)C.[Na+].[Na+]

Specifications

UN Number 3077
Formula Weight 830.81
Quantity 25 g
Physical Form Crystal/Powder
Chemical Name or Material Acid Red 114
EINECSNumber (5)-1518
RTECSNumber QJ6475500
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.