Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acidic Calponin Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP151972
Description
Acidic Calponin Polyclonal specifically detects Acidic Calponin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
Acidic Calponin | |
Polyclonal | |
Western Blot 0.2 - 0.6 ug/ml, Immunohistochemistry 5 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Peptide ELISA Detection limit 1:64000 | |
Q15417 | |
CNN3 | |
Peptide with sequence C-HGEYQDDYPRDYQYS corresponding to C-Terminus according to NP_001830.1. | |
Affinity Purified | |
RUO | |
1266 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
calponin 3, acidic, Calponin, acidic isoform, calponin-3, dJ639P13.2.2 (acidic calponin 3) | |
Goat | |
36 kDa | |
0.1 mg | |
Primary | |
Human. Expected from sequence similarity: Cow. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction