Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acidic Cytokeratin Mouse anti-Human, Clone: KRTL/1377, Abnova™

Catalog No. 89035120 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-120 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-120 Supplier Abnova Corporation Supplier No. MAB15100

Mouse Monoclonal Antibody

Mouse monoclonal antibody raised against partial recombinant human Acidic Cytokeratin.

  • High specificity
  • High reactivity

Specifications

Antigen Acidic Cytokeratin
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone KRTL/1377
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant human Acidic Cytokeratin.
Dilution Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by
Formulation In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
Gene Accession No. P13645;P02533;P19012;P08779;P08727
Gene Alias CK10/K10/KPP
Gene Symbols KRT10
Host Species Mouse
Immunogen Recombinant protein corresponding to human Acidic Cytokeratin.
Purification Method Protein A/G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3858/3861/3866/3868/3880
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.