Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acidic Cytokeratin Rat anti-Human, Clone: OTI8F5, liquid, TrueMAB™
SDP

Catalog No. 501671247
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-1247 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-1247 Supplier OriGene Supplier No. TA190168
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Synthetic peptide sequence is EKVTMQNLND RLASYLDKVR ALEEAN

Type 1 acidic cytokeratin family belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. Mutations in this gene are associated with epidermolytic hyperkeratosis. This gene is located within a cluster of keratin family members on chromosome 17q21.

Specifications

Antigen Acidic Cytokeratin
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OTI8F5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene KRT2
Gene Accession No. P05787, P12035, P13647, P04264, P19013, P02538, P08729, P35908
Gene Alias Acidic CK
Gene Symbols KRT1, KRT2, KRT3, KRT4, KRT5, KRT6A, KRT7, KRT8
Host Species Rat
Immunogen Synthetic peptide corresponding to the conserved region of human type I (Acidic) keratins (including KRT9, KRT10, KRT12, KRT13, KRT14, KRT15, KRT16, KRT17, KRT18, KRT19, KRT20).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3848, 3851, 3850, 3853, 3852, 3849, 3855, 3856
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.