Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ ACINETOBACTER BAUMANNII DERIVED FROM NCTC 13304
SDP

Catalog No. p-7144718
Click to view available options
Quantity:
2 KWIK-STIK
6 KWIK-STIKS
This item is not returnable. View return policy
This item is not returnable. View return policy

Simple, all-inclusive design making life easier for lab techs while reducing chances for errors. Each KWIK-STIK contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid and inoculating swab.

  • Self-contained units of a single organism.
  • Available in a pack of 2 or 6, KWIK-STIKs are universally used for all types of microbiological control testing.

Specifications

ATCC® NCTC 13304
Biosafety Level 2
Certifications/Compliance Clinical Laboratory Standards Institute (CLSI)
Content And Storage 2°C to 8°C
Format KWIK-STIK 2 Pack
Manufacturing Origin United States
Product Type KWIK-STIK
For Use With (Application) Antimicrobial Susceptibility Testing, Microbial Identification
Packaging Type Plastic Canister
Quantity 2 KWIK-STIK
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.