Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Acinetobacter ompA Monoclonal Antibody (5202)

Catalog No. PIMA547562
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547562 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547562 Supplier Invitrogen™ Supplier No. MA547562

Mouse Monoclonal Antibody

Specificity: Specific for the OmpA antigen of several Acinetobacter species. This antibody is reactive with: A. baumannii (4 strains), A. haemolyticus, A. lwoffii (2 strains), Morexella catarrhalis (2 strains). This antibody is non-reactive with: E. coli (mix), Klebsiella pneumoniae, Proteus sp., Pseudomonas aeruginosa, E. faecium, E. fecalis, Group B strep, Staph. saprophyticus, N. gonorrhoeae, Candida sp. Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

ompA acts as a fimbriae subunit.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acinetobacter ompA
Applications ELISA
Classification Monoclonal
Clone 5202
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene ompA
Gene Accession No. P0A911
Gene Alias Fimbrin; ompP5; Outer membrane protein P5
Gene Symbols ompA
Host Species Mouse
Immunogen Proprietary sequence.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 917123
Target Species Bacteria
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.