Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Acinus Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176856
Description
Acinus Polyclonal specifically detects Acinus in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Acinus | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
Acinus, ACN, apoptotic chromatin condensation inducer 1, apoptotic chromatin condensation inducer in the nucleus, fSAP152, functional spliceosome-associated protein 152, KIAA0670DKFZp667N107 | |
Rabbit | |
220 kDa | |
0.1 mg | |
Apoptosis, Cancer, Chromatin Research, Tumor Suppressors | |
22985 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
1 mg/ml | |
Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 0.5-20 μg/mL | |
AAD56724 | |
ACIN1 | |
Antibody was raised against a peptide corresponding to amino acids near the carbosy terminus of human AcinusL, which are identical to those of mouse Acinus. The immunogen is located within the last 50 amino acids of Acinus. Amino Acid Squence: SRSTPVRDRGGRR | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction