Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acinus Polyclonal Antibody, Invitrogen™

Catalog No. PI720313
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI720313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI720313 Supplier Thermo Scientific Supplier No. 720313

Rabbit Polyclonal Antibody

This antibody is predicted to react with Monkey, Bovine, Sheep

ACINUS (apoptotic chromatin condensation inducer in the nucleus) is an induced of chromatin condensation. ACINUS is cleaved by caspase-3 and an additional unknown protease generating a small active peptide, p17, which causes chromatin condensation in vitro when it is added to purified nuclei. ACINUS also induces apoptotic chromatin condensation in cells. ACINUS is ubiquitously expressed. Three different spliced forms of Acinus have been identified in human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acinus
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 30% glycerol and 0.09% sodium azide; pH 7.4
Gene ACIN1
Gene Accession No. Q9UKV3
Gene Alias 2610036I19Rik; 2610510L13Rik; Acin1; ACINUS; acinusL; acinusS; Acn; apoptotic chromatin condensation inducer 1; apoptotic chromatin condensation inducer in the nucleus; C79325; DKFZp667N107; fSAP152; functional spliceosome-associated protein 152; KIAA0670; mKIAA0670
Gene Symbols ACIN1
Host Species Rabbit
Immunogen Peptides corresponding to human ACINUS [1) aa1085-aa1099 2) aa1323-aa1341]
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.