Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Acitretin

Catalog No. AC466440010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466440010 1 g
AC466440050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466440010 Supplier Thermo Scientific Chemicals Supplier No. 466440010
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55079-83-9
Molecular Formula C21H26O3

Specifications

Quantity 1 g
Melting Point 223.0°C to 230.0°C
Formula Weight 326.43
InChI Key IHUNBGSDBOWDMA-AQFIFDHZSA-N
SMILES COC1=CC(C)=C(\C=C\C(\C)=C\C=C\C(\C)=C\C(O)=O)C(C)=C1C
IUPAC Name (2E,4E,6E,8E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
Molecular Weight (g/mol) 326.44
Chemical Name or Material Acitretin
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May damage fertility or the unborn child.

Very toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If skin irritation occurs: Get medical advice/attention.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

EINECSNumber 259-474-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.