Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acitretin 98.0+%, TCI America™
SDP

Catalog No. A3097250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3097250MG 250 mg
A309750MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A3097250MG Supplier TCI America Supplier No. A3097250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 55079-83-9
Molecular Formula C21H26O3
Molecular Weight (g/mol) 326.44
MDL Number MFCD00866632
InChI Key IHUNBGSDBOWDMA-AQFIFDHZSA-N
Synonym (2E,4E,6E,8E)-9-(4-Methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic Acid
PubChem CID 5284513
ChEBI CHEBI:50173
IUPAC Name (2E,4E,6E,8E)-9-(4-methoxy-2,3,6-trimethylphenyl)-3,7-dimethylnona-2,4,6,8-tetraenoic acid
SMILES COC1=CC(C)=C(\C=C\C(\C)=C\C=C\C(\C)=C\C(O)=O)C(C)=C1C

Specifications

Melting Point 230°C
Quantity 250 mg
UN Number 3077
Formula Weight 326.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acitretin
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.