Learn More
Promega ACK Kinase Enzyme System, 1mg
Catalog No. PRV6270 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6270 1 mg
1 options
Catalog No. PRV6270 Supplier Promega Supplier No. V6270
Add to Cart
Add to Cart
Request a Quote
ACK is a tyrosine kinase that binds CDC42Hs in its GTP-bound form and inhibits both the intrinsic and GTPase-activating protein (GAP)-stimulated GTPase activity of CDC42Hs.
  • Easily Screen and Profile ACK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C