Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ACK1 Antibody (ACK 1F4/9), Novus Biologicals™

Mouse Monoclonal Antibody
$173.10
Specifications
Antigen | ACK1 |
---|---|
Clone | ACK 1F4/9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 |
Applications | Western Blot, ELISA |
Description
ACK1 Monoclonal antibody specifically detects ACK1 in Drosophila samples. It is validated for Western Blot, ELISASpecifications
ACK1 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Protein Kinase, Signal Transduction | |
PBS | |
10188 | |
IgM | |
Thiophilic Chromatography |
ACK 1F4/9 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Drosophila | |
ACK-1, ACK1ACKp21cdc42Hs, activated CDC42 kinase 1, activated Cdc42-associated kinase 1, activated p21cdc42Hs kinase, EC 2.7.10, EC 2.7.10.2, FLJ44758, FLJ45547, Tyrosine kinase non-receptor protein 2, tyrosine kinase, non-receptor, 2 | |
ACK peptides (drosophila) | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title