Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ACK1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5144974
Description
Immunogen sequence: PQDIYNVMVQ CWAHKPEDRP TFVALRDFLL EAQPTDMRAL QDFEEPDKLH IQMNDVITVI EGRAENYWWR GQNTRTLCVG PFPRNVVTSV AGLSAQDISQ PLQNSFIHTG HG Antigen sequence identity: Mouse 100%; Rat 100%.
TNK2 (ACK) (activated Cdc42-associated kinase) is a non-receptor tyrosine kinase that signals downstream of Cdc42. TNK2 is known to be involved in tumorigenesis and metastasis, particularly in prostate cancer. Downstream effector of CDC42 which mediates CDC42-dependent cell migration via phosphorylation of BCAR1. May be involved both in adult synaptic function and plasticity and in brain development. Activates AKT1 by phosphorylating it on 'Tyr-176'. Phosphorylates AR on 'Tyr-267' and 'Tyr-363' thereby promoting its recruitment to androgen-responsive enhancers (AREs). Phosphorylates WWOX on 'Tyr-287'. Phosphorylates MCF2, thereby enhancing its activity as a guanine nucleotide exchange factor (GEF) toward Rho family proteins. Contributes to the control of AXL receptor levels. Confers metastatic properties on cancer cells and promotes tumor growth by negatively regulating tumor suppressor such as WWOX and positively regulating pro-survival factors such as AKT1 and AR. Phosphorylates WASP.
Specifications
ACK1 | |
Polyclonal | |
Unconjugated | |
TNK2 | |
ACK; ACK1; Ack-1; Activated CDC42 kinase 1; activated Cdc42-associated kinase 1; activated p21cdc42Hs kinase; Cdc42 GTPase-inhibiting protein; Cdgip; Non-receptor protein tyrosine kinase Ack; p21cdc42Hs; P21cdc42Hs kinase; Pyk1; Tnk2; tnk2 {ECO:0000250; tyrosine kinase non receptor 2; tyrosine kinase non-receptor protein 2; tyrosine kinase, non-receptor, 2; UniProtKB:Q07912} | |
Rabbit | |
Affinity chromatography | |
RUO | |
10188 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid | |
IgG |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q07912 | |
TNK2 | |
Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 357-468. Recombinant protein control fragment (Product #RP-110029). | |
100 μL | |
Primary | |
Human | |
Antibody | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction